Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045745.1 | NP_001039210.1 | 3 - 4 | 386 | 83 | |
XM_012968564.2 | XP_012824018.1 | 4 - 5 | 495 | 83 | ||
XM_018096295.1 | XP_017951784.1 | 3 - 4 | 345 | 83 | ||
GenBank mRNA | CR942488.2 | - | 3 - 4 | 386 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |