Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045745.1 | NP_001039210.1 | 8 - 9 | 1146 | 96 | |
XM_012968564.2 | XP_012824018.1 | 9 - 10 | 1255 | 96 | ||
XM_012968566.2 | XP_012824020.1 | 6 - 7 | 1028 | 96 | ||
XM_018096295.1 | XP_017951784.1 | 8 - 9 | 1105 | 96 | ||
GenBank mRNA | CR942488.2 | - | 8 - 9 | 1146 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Xenopus tropicalis Probe spans exons |