Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mcm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045766.1 | NP_001039231.1 | 16 - 17 | 2292 | 62 | |
GenBank mRNA | BC167938.1 | - | 16 - 17 | 2283 | 62 | |
CR942676.2 | - | 16 - 17 | 2292 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |