Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045770.1 | NP_001039235.1 | 8 - 9 | 851 | 70 | |
GenBank mRNA | BC157333.1 | - | 8 - 9 | 885 | 70 | |
CR942713.1 | - | 8 - 9 | 851 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |