Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: f8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078703.1 | NP_001072171.1 | 10 - 11 | 883 | 98 | |
GenBank mRNA | BC091084.1 | - | 10 - 11 | 813 | 98 | |
BC122015.1 | - | 10 - 11 | 883 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |