Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spata6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078764.1 | NP_001072232.1 | 11 - 12 | 1406 | 120 | |
XM_012964987.2 | XP_012820441.1 | 10 - 11 | 1146 | 120 | ||
XM_018094302.1 | XP_017949791.1 | 12 - 13 | 1475 | 120 | ||
XM_018094304.1 | XP_017949793.1 | 11 - 12 | 1323 | 120 | ||
XM_018094306.1 | XP_017949795.1 | 11 - 12 | 1333 | 120 | ||
XM_018094308.1 | XP_017949797.1 | 10 - 11 | 1294 | 120 | ||
XM_018094309.1 | XP_017949798.1 | 10 - 11 | 1259 | 120 | ||
GenBank mRNA | BC118818.1 | - | 11 - 12 | 1406 | 120 | |
CR760075.2 | - | 11 - 12 | 1230 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |