Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itpkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078788.1 | NP_001072256.1 | 5 - 6 | 1412 | 120 | |
GenBank mRNA | BC121240.1 | - | 5 - 6 | 1412 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |