Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078850.1 | NP_001072318.1 | 2 - 3 | 488 | 103 | |
GenBank mRNA | BC121380.1 | - | 2 - 3 | 488 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |