Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spaca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078914.2 | NP_001072382.2 | 3 - 4 | 402 | 73 | |
XM_012968609.2 | XP_012824063.1 | 3 - 4 | 445 | 73 | ||
GenBank mRNA | BC121496.1 | - | 3 - 4 | 73 | ||
CX886707.1 | - | 3 - 4 | 402 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |