Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spaca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078914.2 | NP_001072382.2 | 4 - 5 | 561 | 59 | |
GenBank mRNA | BC121496.1 | - | 4 - 5 | 558 | 59 | |
CX886707.1 | - | 4 - 5 | 561 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |