Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tesc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078955.1 | NP_001072423.1 | 5 - 6 | 731 | 120 | |
GenBank mRNA | BC121662.1 | - | 5 - 6 | 731 | 120 | |
BC135886.1 | - | 5 - 6 | 583 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |