Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079216.1 | NP_001072684.1 | 7 - 8 | 1364 | 91 | |
XM_018097781.1 | XP_017953270.1 | 7 - 8 | 1301 | 91 | ||
GenBank mRNA | BC124016.1 | - | 7 - 8 | 1364 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |