Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079219.1 | NP_001072687.1 | 10 - 11 | 1113 | 98 | |
XM_012965802.1 | XP_012821256.1 | 10 - 11 | 1120 | 98 | ||
XM_018094605.1 | XP_017950094.1 | 10 - 11 | 814 | 98 | ||
GenBank mRNA | BC124020.1 | - | 10 - 11 | 1113 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |