Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079219.1 | NP_001072687.1 | 11 - 12 | 1240 | 103 | |
XM_012965802.1 | XP_012821256.1 | 11 - 12 | 1247 | 103 | ||
XM_018094605.1 | XP_017950094.1 | 11 - 12 | 941 | 103 | ||
GenBank mRNA | BC124020.1 | - | 11 - 12 | 1240 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |