Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079244.1 | NP_001072712.1 | 6 - 7 | 677 | 108 | |
XM_012965828.2 | XP_012821282.1 | 7 - 8 | 924 | 108 | ||
GenBank mRNA | BC124065.1 | - | 6 - 7 | 677 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |