Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079247.1 | NP_001072715.1 | 11 - 12 | 1590 | 68 | |
GenBank mRNA | BC125674.1 | - | 11 - 12 | 1590 | 68 | |
CT025320.2 | - | 11 - 12 | 1588 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |