Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shroom4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079276.1 | NP_001072744.1 | 8 - 9 | 4724 | 81 | |
XM_018096333.1 | XP_017951822.1 | 9 - 10 | 4877 | 81 | ||
XM_018096334.1 | XP_017951823.1 | 10 - 11 | 4943 | 81 | ||
GenBank mRNA | BC125736.1 | - | 8 - 9 | 4724 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |