Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cybc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079369.1 | NP_001072837.1 | 5 - 6 | 340 | 75 | |
GenBank mRNA | BC121946.1 | - | 5 - 6 | 340 | 75 | |
CT025438.2 | - | 5 - 6 | 349 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |