Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079374.1 | NP_001072842.1 | 4 - 5 | 726 | 95 | |
GenBank mRNA | BC121978.1 | - | 4 - 5 | 726 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |