Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: neto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079444.1 | NP_001072912.1 | 2 - 3 | 428 | 62 | |
GenBank mRNA | BC121922.1 | - | 2 - 3 | 428 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |