Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079451.1 | NP_001072919.1 | 12 - 13 | 1000 | 98 | |
XM_012970593.1 | XP_012826047.1 | 12 - 13 | 1234 | 98 | ||
GenBank mRNA | BC121427.1 | - | 12 - 13 | 1000 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |