Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr2c2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016306.3 | NP_001016306.1 | 2 - 3 | 229 | 102 | |
GenBank mRNA | BC129029.1 | - | 2 - 3 | 229 | 102 | |
CR761499.2 | - | 2 - 3 | 279 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |