Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079015.1 | NP_001072483.1 | 1 - 2 | 3010 | 102 | |
XM_012965336.2 | XP_012820790.1 | 1 - 2 | 3043 | 102 | ||
GenBank mRNA | BC121910.1 | - | 1 - 2 | 3010 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |