Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079044.1 | NP_001072512.1 | 11 - 12 | 1362 | 81 | |
GenBank mRNA | BC121953.1 | - | 11 - 12 | 1362 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |