Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079113.1 | NP_001072581.1 | 7 - 8 | 1603 | 83 | |
XM_012961351.2 | XP_012816805.1 | 7 - 8 | 1891 | 83 | ||
XM_012961354.2 | XP_012816808.1 | 7 - 8 | 1890 | 83 | ||
GenBank mRNA | BC122939.1 | - | 7 - 8 | 1603 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |