Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079267.1 | NP_001072735.1 | 8 - 9 | 2766 | 127 | |
GenBank mRNA | BC125715.1 | - | 8 - 9 | 2766 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |