Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079430.1 | NP_001072898.1 | 2 - 3 | 508 | 68 | |
GenBank mRNA | BC118858.1 | - | 2 - 3 | 508 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |