Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097228.1 | NP_001090697.1 | 2 - 3 | 718 | 80 | |
XM_012962527.2 | XP_012817981.1 | 2 - 3 | 854 | 80 | ||
XM_012962528.2 | XP_012817982.1 | 3 - 4 | 681 | 80 | ||
GenBank mRNA | BC128618.1 | - | 2 - 3 | 718 | 80 | |
CU025012.1 | - | 2 - 3 | 747 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |