Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st8sia5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097279.1 | NP_001090748.1 | 3 - 4 | 310 | 96 | |
XM_012956876.2 | XP_012812330.1 | 4 - 5 | 950 | 96 | ||
XM_012956917.2 | XP_012812371.1 | 3 - 4 | 842 | 96 | ||
GenBank mRNA | AM422136.1 | - | 3 - 4 | 310 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |