Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097322.1 | NP_001090791.1 | 5 - 6 | 706 | 109 | |
XM_012965827.2 | XP_012821281.1 | 5 - 6 | 820 | 109 | ||
GenBank mRNA | BC135183.1 | - | 5 - 6 | 706 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |