Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097334.2 | NP_001090803.1 | 3 - 4 | 798 | 97 | |
GenBank mRNA | BC082743.1 | - | 3 - 4 | 787 | 97 | |
CX347149.2 | - | 3 - 4 | 798 | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |