Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097392.1 | NP_001090861.1 | 2 - 3 | 360 | 87 | |
GenBank mRNA | BC135248.2 | - | 2 - 3 | 360 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |