Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100205.1 | NP_001093675.1 | 5 - 6 | 1321 | 64 | |
GenBank mRNA | BC135990.1 | - | 5 - 6 | 1321 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |