Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100208.1 | NP_001093678.1 | 3 - 4 | 1014 | 98 | |
XM_012969938.2 | XP_012825392.1 | 3 - 4 | 985 | 98 | ||
GenBank mRNA | BC136009.1 | - | 3 - 4 | 1014 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |