Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100268.3 | NP_001093738.2 | 7 - 8 | 1267 | 73 | |
GenBank mRNA | BC135422.2 | - | 7 - 8 | 1101 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |