Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zc3h6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102732.2 | NP_001096202.2 | 3 - 4 | 586 | 84 | |
GenBank mRNA | BC135562.1 | - | 3 - 4 | 578 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |