Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kyat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102788.1 | NP_001096258.1 | 4 - 5 | 364 | 73 | |
GenBank mRNA | BC084158.1 | - | 3 - 4 | 237 | 73 | |
BC135439.1 | - | 4 - 5 | 364 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |