Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102789.1 | NP_001096259.1 | 851 | 73 | ||
GenBank mRNA | BC135444.1 | - | 851 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |