Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tcta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102794.2 | NP_001096264.1 | 1 - 2 | 416 | 82 | |
GenBank mRNA | BC135472.1 | - | 1 - 2 | 82 | ||
BC155508.1 | - | 1 - 2 | 305 | 82 | ||
CN094320.1 | - | 1 - 2 | 82 | |||
ES679293.1 | - | 1 - 2 | 410 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |