Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prtfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102892.1 | NP_001096362.1 | 8 - 9 | 605 | 75 | |
XM_018094511.1 | XP_017950000.1 | 8 - 9 | 886 | 75 | ||
XM_018094512.1 | XP_017950001.1 | 7 - 8 | 865 | 75 | ||
GenBank mRNA | BC135868.1 | - | 8 - 9 | 605 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |