Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ormdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102917.1 | NP_001096387.1 | 2 - 3 | 246 | 84 | |
XM_012969979.2 | XP_012825433.1 | 3 - 4 | 325 | 84 | ||
GenBank mRNA | BC135976.1 | - | 2 - 3 | 246 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |