Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102972.3 | NP_001096442.2 | 1 - 2 | 597 | 102 | |
GenBank mRNA | AL899990.2 | - | 1 - 2 | 589 | 102 | |
BC136141.1 | - | 1 - 2 | 597 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |