Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102994.1 | NP_001096464.1 | 8 - 9 | 1415 | 98 | |
GenBank mRNA | BC136195.1 | - | 8 - 9 | 1415 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |