Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016598.3 | NP_001016598.1 | 5 - 6 | 426 | 100 | |
GenBank mRNA | BC158895.1 | - | 5 - 6 | 426 | 100 | |
BC170783.1 | - | 5 - 6 | 394 | 100 | ||
BC170789.1 | - | 5 - 6 | 394 | 100 | ||
CR760568.2 | - | 5 - 6 | 800 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |