Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c1orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016665.3 | NP_001016665.1 | 4 - 5 | 409 | 77 | |
XM_012966617.2 | XP_012822071.1 | 5 - 6 | 593 | 77 | ||
GenBank mRNA | BC158946.1 | - | 4 - 5 | 409 | 77 | |
BC171198.1 | - | 4 - 5 | 400 | 77 | ||
BC171202.1 | - | 4 - 5 | 400 | 77 | ||
CR760289.2 | - | 5 - 6 | 569 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |