Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103083.1 | NP_001096553.1 | 1 - 2 | 95 | 79 | |
GenBank mRNA | DQ658160.1 | - | 1 - 2 | 95 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Xenopus tropicalis Probe spans exons |