Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110049.1 | NP_001103519.1 | 2 - 3 | 456 | 78 | |
XM_012969996.2 | XP_012825450.1 | 2 - 3 | 554 | 78 | ||
GenBank mRNA | BC153745.1 | - | 2 - 3 | 456 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |