Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110050.1 | NP_001103520.2 | 10 - 11 | 1497 | 98 | |
XM_012956327.2 | XP_012811781.1 | 11 - 12 | 2309 | 98 | ||
XM_012956331.2 | XP_012811785.1 | 11 - 12 | 2359 | 98 | ||
XM_012956333.2 | XP_012811787.1 | 11 - 12 | 2306 | 98 | ||
XM_012956334.2 | XP_012811788.1 | 11 - 12 | 2355 | 98 | ||
XM_012956335.2 | XP_012811789.1 | 11 - 12 | 2295 | 98 | ||
XM_018091054.1 | XP_017946543.1 | 11 - 12 | 2310 | 98 | ||
GenBank mRNA | BC153338.1 | - | 10 - 11 | 1497 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |