Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110050.1 | NP_001103520.2 | 11 - 12 | 1662 | 77 | |
XM_012956327.2 | XP_012811781.1 | 12 - 13 | 2474 | 77 | ||
XM_012956331.2 | XP_012811785.1 | 12 - 13 | 2524 | 77 | ||
XM_012956333.2 | XP_012811787.1 | 12 - 13 | 2471 | 77 | ||
XM_012956334.2 | XP_012811788.1 | 12 - 13 | 2520 | 77 | ||
XM_012956335.2 | XP_012811789.1 | 12 - 13 | 2460 | 77 | ||
GenBank mRNA | BC153338.1 | - | 11 - 12 | 1662 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |