Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsnare1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112923.1 | NP_001106394.1 | 9 - 10 | 813 | 84 | |
GenBank mRNA | BC152038.1 | - | 9 - 10 | 813 | 84 | |
CT025175.1 | - | 10 - 11 | 853 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |